Pentafluorophenyl 4-(acetylamino)benzenesulfonate ≥95%

Supplier: Matrix Scientific

043134-500MG 043134-5G
101859-342EA 375.38 USD
101859-342 101859-346
Pentafluorophenyl 4-(acetylamino)benzenesulfonate ≥95%
Pentafluorophenyl 4-(acetylamino)benzenesulphonate

Hazard: Irritant

Formula: C₁₄H₈F₅NO₄S
Melting Pt: 168-170°C
MDL Number: MFCD05975141
CAS Number: 886360-93-6

Order Now

Specification Test Results

Melting point 168-170°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR