{3-[3-(Trifluoromethyl)phenyl]-1,2,4-oxadiazol-5-yl}methanol ≥95%

Supplier: Matrix Scientific

043132-500MG 043132-5G
101859-330EA 747.69 USD
101859-330 101859-334
{3-[3-(Trifluoromethyl)phenyl]-1,2,4-oxadiazol-5-yl}methanol ≥95%
{3-[3-(Trifluoromethyl)phenyl]-1,2,4-oxadiazol-5-yl}methanol

Hazard: Irritant

Formula: C₁₀H₇F₃N₂O₂
Melting Pt: 66…67 °C
MDL Number: MFCD06200920
CAS Number: 886361-32-6

Order Now

Specification Test Results

Melting point 66-67°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR