Pentafluorophenyl 8-quinolinesulfonate ≥95%
Supplier: Matrix Scientific
043129-500MG
043129-5G
101859-318EA
375.38
USD
101859-318
101859-322
Pentafluorophenyl 8-quinolinesulfonate ≥95%
Pentafluorophenyl 8-quinolinesulphonate
Hazard: Irritant
Formula:
C₉H₇NO₃S Melting Pt: 155…157 °C |
MDL Number:
MFCD05975139 CAS Number: 885950-64-1 |
Specification Test Results
Melting point | 155-157°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...