Pentafluorophenyl 2-methyl-5-nitrobenzenesulfonate ≥95%

Supplier: Matrix Scientific

043127-500MG 043127-5G
101859-306EA 375.38 USD
101859-306 101859-310
Pentafluorophenyl 2-methyl-5-nitrobenzenesulfonate ≥95%
Pentafluorophenyl 2-methyl-5-nitrobenzenesulphonate

Hazard: Irritant

Formula: C₁₃H₆F₅NO₅S
Melting Pt: 123…125 °C
MDL Number: MFCD05975137
CAS Number: 885950-60-7

Order Now

Specification Test Results

Melting point 123-125°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR