Methyl-3-methyl-2-nitrobenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 3-methyl-2-nitrobenzoate

043123-1G 043123-25G
101859-286EA 200 USD
101859-286 102551-314
Methyl-3-methyl-2-nitrobenzoate ≥95%
Methyl 3-methyl-2-nitrobenzoate

Hazard: Irritant

Formula: C₉H₉NO₄
MW: 195.17 g/mol
Melting Pt: 72…73 °C
MDL Number: MFCD01003947
CAS Number: 5471-82-9

Order Now

Specification Test Results

Melting point 72-73°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR