Methyl-2-[(4-formyl-1,3-dimethyl-1H-pyrazol-5-yl)sulfanyl]benzenecarboxylate ≥95%
Supplier: Matrix Scientific
043119-500MG
043119-5G
101859-264EA
747.69
USD
101859-264
101859-268
Methyl-2-[(4-formyl-1,3-dimethyl-1H-pyrazol-5-yl)sulfanyl]benzenecarboxylate ≥95%
Methyl-2-[(4-formyl-1,3-dimethyl-1H-pyrazol-5-yl)sulfanyl]benzenecarboxylate
Hazard: Irritant
Formula:
C₁₄H₁₄N₂O₃S Melting Pt: 96…99 °C |
MDL Number:
MFCD04117786 CAS Number: 852700-50-6 |
Specification Test Results
Melting point | 96-99°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...