4-(2-Chloro-6-fluorobenzyl)-3,5-dimethyl-1H-pyrazole ≥95%
Supplier: Matrix Scientific
043105-500MG
043105-5G
101859-186EA
438.46
USD
101859-186
101859-190
4-(2-Chloro-6-fluorobenzyl)-3,5-dimethyl-1H-pyrazole ≥95%
4-(2-Chloro-6-fluorobenzyl)-3,5-dimethyl-1H-pyrazole
Hazard: Irritant
Formula:
C₁₂H₁₂ClFN₂ Melting Pt: 133…134 °C |
MDL Number:
MFCD00120327 CAS Number: 387352-96-7 |
Specification Test Results
Melting point | 133-134°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...