4-[(2-Fluorobenzyl)oxy]-N'-hydroxybenzenecarboximidamide ≥95%

Supplier: Matrix Scientific

043098-500MG 043098-5G
101859-146EA 921.54 USD
101859-146 101859-150
4-[(2-Fluorobenzyl)oxy]-N'-hydroxybenzenecarboximidamide ≥95%
4-[(2-Fluorobenzyl)oxy]-N'-hydroxybenzenecarboximidamide

Hazard: Irritant

Formula: C₁₄H₁₃FN₂O₂
Melting Pt: 138…139 °C
MDL Number: MFCD00662630
CAS Number: 261965-35-9

Order Now

Specification Test Results

Melting point 138-139°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR