4-(1,3-Dioxolan-2-yl)-N'-hydroxybenzenecarboximidamide ≥95%

Supplier: Matrix Scientific

043096-500MG 043096-5G
101859-134EA 921.54 USD
101859-134 101859-138
4-(1,3-Dioxolan-2-yl)-N'-hydroxybenzenecarboximidamide ≥95%
4-(1,3-Dioxolan-2-yl)-N'-hydroxybenzenecarboximidamide

Hazard: Irritant

Formula: C₁₀H₁₂N₂O₃
MW: 208.22 g/mol
Melting Pt: 144…146 °C
MDL Number: MFCD02089742
CAS Number: 852691-00-0

Order Now

Specification Test Results

Melting point 144-146°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR