5-(2-Thienyl)-3-(trifluoromethyl)-1H-pyrazole ≥95%
Supplier: Matrix Scientific
043094-500MG
043094-5G
101859-122EA
921.54
USD
101859-122
101859-126
5-(2-Thienyl)-3-(trifluoromethyl)-1H-pyrazole ≥95%
5-(2-Thienyl)-3-(trifluoromethyl)-1H-pyrazole
Hazard: Irritant
Formula:
C₈H₅F₃N₂S MW: 218.2 g/mol Melting Pt: 120…122 °C |
MDL Number:
MFCD00459838 CAS Number: 128228-96-6 |
Specification Test Results
Melting point | 120-122°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...