[3-(2-Pyrimidinyloxy)phenyl]methanol ≥95%

Supplier: Matrix Scientific

043092-500MG 043092-5G
101859-110EA 278.46 USD
101859-110 101859-114
[3-(2-Pyrimidinyloxy)phenyl]methanol ≥95%
[3-(2-Pyrimidinyloxy)phenyl]methanol

Hazard: Irritant

Formula: C₁₁H₁₀N₂O₂
MW: 202.21 g/mol
Melting Pt: 147…150 °C
MDL Number: MFCD03791155
CAS Number: 82440-25-3

Order Now

Specification Test Results

Melting point 147-150°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR