4-[(2-Fluorobenzyl)oxy]benzenecarbonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(2-Fluorobenzyloxy)benzonitrile

043090-500MG 043090-5G
101859-098EA 181.54 USD
101859-098 101859-102
4-[(2-Fluorobenzyl)oxy]benzenecarbonitrile ≥95%
4-[(2-Fluorobenzyl)oxy]benzenecarbonitrile

Hazard: Irritant

Formula: C₁₄H₁₀FNO
MW: 227.24 g/mol
Melting Pt: 80…81 °C
MDL Number: MFCD02681655
CAS Number: 852690-97-2

Order Now

Specification Test Results

Melting point 80-81°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR