5-Cyclopropyl-2H-1,2,3,4-tetraazole ≥97%

Supplier: Matrix Scientific

043082-500MG 043082-5G
101859-062EA 512.31 USD
101859-062 101859-066
5-Cyclopropyl-2H-1,2,3,4-tetraazole ≥97%
5-Cyclopropyl-2H-1,2,3,4-tetraazole

Hazard: Irritant

Formula: C₄H₆N₄
Melting Pt: 152…154 °C
MDL Number: MFCD00221169
CAS Number: 27943-07-3

Order Now

Specification Test Results

Melting point 152-154°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR