2-[4-(4-Chlorophenyl)-1,3-thiazol-2-yl]acetonitrile ≥95%

Supplier: Matrix Scientific

043079-500MG 043079-5G
101859-050EA 181.54 USD
101859-050 101859-054
2-[4-(4-Chlorophenyl)-1,3-thiazol-2-yl]acetonitrile ≥95%
2-[4-(4-Chlorophenyl)-1,3-thiazol-2-yl]acetonitrile

Hazard: Irritant

Formula: C₁₁H₇ClN₂S
MW: 234.71 g/mol
Melting Pt: 124…126 °C
MDL Number: MFCD00052875
CAS Number: 17969-48-1

Order Now

Specification Test Results

Melting point 124-126°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR