Methyl-4-acetyl-1H-pyrrole-2-carboxylate ≥95%

Supplier: Matrix Scientific

043070-500MG 043070-5G
101858-998EA 747.69 USD
101858-998 101859-002
Methyl-4-acetyl-1H-pyrrole-2-carboxylate ≥95%
Methyl-4-acetyl-1H-pyrrole-2-carboxylate

Hazard: Irritant

Formula: C₈H₉NO₃
MW: 167.17 g/mol
Melting Pt: 118…120 °C
MDL Number: MFCD02571735
CAS Number: 40611-82-3

Order Now

Specification Test Results

Melting point 118-120°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR