3-[4-Methoxy-3-(2-propynyloxy)phenyl]acrylic acid ≥95%

Supplier: Matrix Scientific

043068-500MG 043068-5G
101858-986EA 375.38 USD
101858-986 101858-990
3-[4-Methoxy-3-(2-propynyloxy)phenyl]acrylic acid ≥95%
3-[4-Methoxy-3-(2-propynyloxy)phenyl]acrylic acid

Hazard: Irritant

Formula: C₁₃H₁₂O₄
Melting Pt: 210…213 °C
MDL Number: MFCD03001239
CAS Number: 385383-51-7

Order Now

Specification Test Results

Melting point 210-213°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR