Methyl-2-[(3-fluorobenzyl)sulfanyl]benzenecarboxylate ≥95%

Supplier: Matrix Scientific

043054-500MG 043054-5G
101858-910EA 278.46 USD
101858-910 101858-914
Methyl-2-[(3-fluorobenzyl)sulfanyl]benzenecarboxylate ≥95%
Methyl-2-[(3-fluorobenzyl)sulfanyl]benzenecarboxylate

Hazard: Irritant

Formula: C₁₅H₁₃FO₂S
MW: 276.33 g/mol
Melting Pt: 70…72 °C
MDL Number: MFCD03001233
CAS Number: 385383-54-0

Order Now

Specification Test Results

Melting point 70-72°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR