Methyl-2-[(2,4-dichlorobenzyl)sulfanyl]benzenecarboxylate ≥95%

Supplier: Matrix Scientific

043052-500MG 043052-5G
101858-898EA 278.46 USD
101858-898 101858-902
Methyl-2-[(2,4-dichlorobenzyl)sulfanyl]benzenecarboxylate ≥95%
Methyl-2-[(2,4-dichlorobenzyl)sulfanyl]benzenecarboxylate

Hazard: Irritant

Formula: C₁₅H₁₂Cl₂O₂S
Melting Pt: 114…117 °C
MDL Number: MFCD03001231
CAS Number: 308851-38-9

Order Now

Specification Test Results

Melting point 114-117°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR