1-Benzyl-3,5-dibromo-1H-1,2,4-triazole ≥95%
Supplier: Matrix Scientific
043025-500MG
043025-5G
101858-770EA
207.69
USD
101858-770
101858-774
1-Benzyl-3,5-dibromo-1H-1,2,4-triazole ≥95%
1-Benzyl-3,5-dibromo-1H-1,2,4-triazole
Hazard: Irritant
Formula:
C₉H₇Br₂N₃ Melting Pt: 69…70 °C |
MDL Number:
MFCD01568839 CAS Number: 106724-85-0 |
Specification Test Results
Melting point | 69-70°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...