Methyl-4-[(2-methoxy-2-oxoethyl)sulfanyl]-3-nitrobenzenecarboxylate ≥95%

Supplier: Matrix Scientific

043018-500MG 043018-5G
101858-730EA 181.54 USD
101858-730 101858-734
Methyl-4-[(2-methoxy-2-oxoethyl)sulfanyl]-3-nitrobenzenecarboxylate ≥95%
Methyl-4-[(2-methoxy-2-oxoethyl)sulfanyl]-3-nitrobenzenecarboxylate

Hazard: Irritant

Formula: C₁₁H₁₁NO₆S
Melting Pt: 124…126 °C
MDL Number: MFCD00187196
CAS Number: 202131-27-9

Order Now

Specification Test Results

Melting point 124-126°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR