2-(4-Phenylpiperazino)-1,3-thiazole-5-carbaldehyde ≥95%

Supplier: Matrix Scientific

043016-500MG 043016-5G
101858-718EA 1296.92 USD
101858-718 101858-722
2-(4-Phenylpiperazino)-1,3-thiazole-5-carbaldehyde ≥95%
2-(4-Phenylpiperazino)-1,3-thiazole-5-carbaldehyde

Hazard: Irritant

Formula: C₁₄H₁₅N₃OS
MW: 273.36 g/mol
Melting Pt: 118…119 °C
MDL Number: MFCD01568830
CAS Number: 303987-39-5

Order Now

Specification Test Results

Melting point 118-119°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR