2-({Imino[3-(trifluoromethyl)phenyl]methyl}amino)acetic acid ≥95%

Supplier: Matrix Scientific

Synonyms: N-(2-Carboxymethyl)-3-(trifluoromethyl)benzamidine

043007-500MG 043007-5G
101858-682EA 484.62 USD
101858-682 101858-686
2-({Imino[3-(trifluoromethyl)phenyl]methyl}amino)acetic acid ≥95%
2-({Imino[3-(trifluoromethyl)phenyl]methyl}amino)acetic acid

Hazard: Irritant

Formula: C₁₀H₉F₃N₂O₂
MW: 246.19 g/mol
Melting Pt: 204…208 °C
MDL Number: MFCD01935986
CAS Number: 338982-13-1

Order Now

Specification Test Results

Melting point 204-208°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR