Ethyl-1-methyl-3-trifluoromethyl-1H-pyrazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxylate

043005-500MG 043005-5G
101858-670EA 24.62 USD
101858-670 101858-674
Ethyl-1-methyl-3-trifluoromethyl-1H-pyrazole-4-carboxylate ≥95%
Ethyl-1-methyl-3-trifluoromethyl-1H-pyrazole-4-carboxylate

Hazard: Irritant

Formula: C₈H₉F₃N₂O₂
MW: 222.167 g/mol
Melting Pt: 60…61 °C
MDL Number: MFCD02083398
CAS Number: 111493-74-4

Order Now

Specification Test Results

Melting point 60-61°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR