3-Chloro-2,2-dimethyl-N-(3-nitrophenyl)propanamide ≥95%

Supplier: Matrix Scientific

043001-500MG 043001-5G
101858-654EA 181.54 USD
101858-654 101858-658
3-Chloro-2,2-dimethyl-N-(3-nitrophenyl)propanamide ≥95%
3-Chloro-2,2-dimethyl-N-(3-nitrophenyl)propanamide

Hazard: Irritant

Formula: C₁₁H₁₃ClN₂O₃
Melting Pt: 115…117 °C
MDL Number: MFCD00793375
CAS Number: 303985-82-2

Order Now

Specification Test Results

Melting point 115-117°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR