1-[3-(4-Chlorophenyl)-5-isoxazolyl]-1-ethanone ≥95%
Supplier: Matrix Scientific
042999-500MG
042999-5G
101858-642EA
556.92
USD
101858-642
101858-646
1-[3-(4-Chlorophenyl)-5-isoxazolyl]-1-ethanone ≥95%
1-[3-(4-Chlorophenyl)-5-isoxazolyl]-1-ethanone
Hazard: Irritant
Formula:
C₁₁H₈ClNO₂ MW: 221.64 g/mol Melting Pt: 148…149 °C |
MDL Number:
MFCD00662755 CAS Number: 37091-33-1 |
Specification Test Results
Melting point | 148-149°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...