[3-(4-Methoxyphenyl)-5-isoxazolyl]methanol ≥95%

Supplier: Matrix Scientific

042989-500MG 042989-5G
101858-602EA 375.38 USD
101858-602 101858-606
[3-(4-Methoxyphenyl)-5-isoxazolyl]methanol ≥95%
[3-(4-Methoxyphenyl)-5-isoxazolyl]methanol

Hazard: Irritant

Formula: C₁₁H₁₁NO₃
Melting Pt: 97…98 °C
MDL Number: MFCD04124287
CAS Number: 206055-86-9

Order Now

Specification Test Results

Melting point 97-98°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR