3-(1,2,3-Thiadiazol-4-yl)benzenol ≥95%

Supplier: Matrix Scientific

042985-500MG 042985-5G
101858-582EA 278.46 USD
101858-582 101858-586
3-(1,2,3-Thiadiazol-4-yl)benzenol ≥95%
3-(1,2,3-Thiadiazol-4-yl)benzenol

Hazard: Irritant

Formula: C₈H₆N₂OS
Melting Pt: 140…142 °C
MDL Number: MFCD00792977
CAS Number: 303149-00-0

Order Now

Specification Test Results

Melting point 140-142°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR