5-Amino-2-[(4-methylphenyl)sulfanyl]benzenecarbonitrile ≥95%
Supplier: Matrix Scientific
042977-500MG
042977-5G
101858-542EA
375.38
USD
101858-542
101858-546
5-Amino-2-[(4-methylphenyl)sulfanyl]benzenecarbonitrile ≥95%
5-Amino-2-[(4-methylphenyl)sulfanyl]benzenecarbonitrile
Hazard: Irritant
Formula:
C₁₄H₁₂N₂S MW: 240.33 g/mol Melting Pt: 93…96 °C |
MDL Number:
MFCD00793509 CAS Number: 303148-03-0 |
Specification Test Results
Melting point | 93-96°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...