6-[(Phenylsulfanyl)methyl]-2-(3-pyridinyl)-4-pyrimidinol ≥95%

Supplier: Matrix Scientific

042975-500MG 042975-5G
101858-530EA 747.69 USD
101858-530 101858-534
6-[(Phenylsulfanyl)methyl]-2-(3-pyridinyl)-4-pyrimidinol ≥95%
6-[(Phenylsulfanyl)methyl]-2-(3-pyridinyl)-4-pyrimidinol

Hazard: Irritant

Formula: C₁₆H₁₃N₃OS
Melting Pt: 180…182 °C
MDL Number: MFCD06200916
CAS Number: 852377-18-5

Order Now

Specification Test Results

Melting point 180-182°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR