7-(2-Pyridinyl)[1,2,4]triazolo[1,5-a]pyrimidin-2-amine ≥95%

Supplier: Matrix Scientific

042971-500MG 042971-5G
101858-506EA 269.23 USD
101858-506 101858-510
7-(2-Pyridinyl)[1,2,4]triazolo[1,5-a]pyrimidin-2-amine ≥95%
7-(2-Pyridinyl)[1,2,4]triazolo[1,5-a]pyrimidin-2-amine

Hazard: Irritant

Formula: C₁₀H₈N₆
Melting Pt: 252…254 °C
MDL Number: MFCD00232454
CAS Number: 303145-68-8

Order Now

Specification Test Results

Melting point 252-254°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR