2-[5-(2-Fluorobenzoyl)-2-thienyl]acetonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(Cyanomethyl)-5-(2-fluorobenzoyl)thiophene

042965-500MG 042965-5G
101858-472EA 278.46 USD
101858-472 101858-476
2-[5-(2-Fluorobenzoyl)-2-thienyl]acetonitrile ≥95%
2-[5-(2-Fluorobenzoyl)-2-thienyl]acetonitrile

Hazard: Irritant

Formula: C₁₃H₈FNOS
MW: 245.28 g/mol
Melting Pt: 84…86 °C
MDL Number: MFCD00664551
CAS Number: 303144-50-5

Order Now

Specification Test Results

Melting point 84-86°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR