Methyl-3-[(5-nitro-2-pyridinyl)oxy]-2-thiophenecarboxylate ≥95%
Supplier: Matrix Scientific
042956-500MG
042956-5G
101858-424EA
556.92
USD
101858-424
101858-428
Methyl-3-[(5-nitro-2-pyridinyl)oxy]-2-thiophenecarboxylate ≥95%
Methyl-3-[(5-nitro-2-pyridinyl)oxy]-2-thiophenecarboxylate
Hazard: Irritant
Formula:
C₁₁H₈N₂O₅S Melting Pt: 118…120 °C |
MDL Number:
MFCD00214990 CAS Number: 303152-15-0 |
Specification Test Results
Melting point | 118-120°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...