4-(4-Fluorophenyl)-6-hydroxy-2-phenyl-5-pyrimidinecarbonitrile ≥95%
Supplier: Matrix Scientific
042954-500MG
042954-5G
101858-412EA
130.77
USD
101858-412
101858-416
4-(4-Fluorophenyl)-6-hydroxy-2-phenyl-5-pyrimidinecarbonitrile ≥95%
4-(4-Fluorophenyl)-6-hydroxy-2-phenyl-5-pyrimidinecarbonitrile
Hazard: Irritant
Formula:
C₁₇H₁₀FN₃O MW: 291.29 g/mol Melting Pt: 330 °C |
MDL Number:
MFCD00794103 CAS Number: 477853-95-5 |
Specification Test Results
Melting point | 330°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...