3-{[3-(Trifluoromethyl)benzyl]sulfanyl}-1H-1,2,4-triazol-5-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Amino-3-[3-(trifluoromethyl)benzylthio]-1H-1,2,4-triazole

042947-500MG 042947-5G
101858-380EA 375.38 USD
101858-380 101858-384
3-{[3-(Trifluoromethyl)benzyl]sulfanyl}-1H-1,2,4-triazol-5-amine ≥95%
3-{[3-(Trifluoromethyl)benzyl]sulfanyl}-1H-1,2,4-triazol-5-amine

Hazard: Irritant

Formula: C₁₀H₉F₃N₄S
MW: 274.27 g/mol
Melting Pt: 136…138 °C
MDL Number: MFCD00141527
CAS Number: 303150-56-3

Order Now

Specification Test Results

Melting point 136-138°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR