2-(Phenylsulfonyl)-1,4-benzenediol ≥95%

Supplier: Matrix Scientific

042945-500MG 042945-5G
101858-368EA 278.46 USD
101858-368 101858-372
2-(Phenylsulfonyl)-1,4-benzenediol ≥95%
2-(Phenylsulfonyl)-1,4-benzenediol

Hazard: Irritant

Formula: C₁₂H₁₀O₄S
MW: 250.28 g/mol
Melting Pt: 198…200 °C
MDL Number: MFCD00159099
CAS Number: 23156-75-4

Order Now

Specification Test Results

Melting point 198-200°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR