3-[(2,4-Dichlorobenzyl)sulfanyl]-1H-1,2,4-triazol-5-amine ≥95%

Supplier: Matrix Scientific

042941-500MG 042941-5G
101858-344EA 67.69 USD
101858-344 101858-348
3-[(2,4-Dichlorobenzyl)sulfanyl]-1H-1,2,4-triazol-5-amine ≥95%
3-[(2,4-Dichlorobenzyl)sulfanyl]-1H-1,2,4-triazol-5-amine

Hazard: Irritant

Formula: C₉H₈Cl₂N₄S
Melting Pt: 156…158 °C
MDL Number: MFCD00141525
CAS Number: 303150-34-7

Order Now

Specification Test Results

Melting point 156-158°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR