1-[4-(Chloromethyl)-1,3-thiazol-2-yl]-4-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]piperazine ≥95%

Supplier: Matrix Scientific

042937-500MG 042937-5G
101858-320EA 1483.08 USD
101858-320 101858-324
1-[4-(Chloromethyl)-1,3-thiazol-2-yl]-4-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]piperazine ≥95%
1-[4-(Chloromethyl)-1,3-thiazol-2-yl]-4-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]piperazine

Hazard: Irritant

Formula: C₁₄H₁₃Cl₂F₃N₄S
MW: 397.25 g/mol
Melting Pt: 97…99 °C
MDL Number: MFCD00172020
CAS Number: 303150-21-2

Order Now

Specification Test Results

Melting point 97-99°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR