1-(6-Chloro-3-pyridazinyl)-4-piperidinecarboxamide ≥95%
Supplier: Matrix Scientific
Synonyms:
1-(6-Chloro-3-pyridazinyl)piperidine-4-carboxamide
042931-500MG
042931-5G
101858-288EA
556.92
USD
101858-288
101858-292
1-(6-Chloro-3-pyridazinyl)-4-piperidinecarboxamide ≥95%
1-(6-Chloro-3-pyridazinyl)-4-piperidinecarboxamide
Hazard: Irritant
Formula:
C₁₀H₁₃ClN₄O MW: 240.69 g/mol Melting Pt: 216…218 °C |
MDL Number:
MFCD00180782 CAS Number: 303149-97-5 |
Specification Test Results
Melting point | 216-218°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...