5-(4-Chlorophenoxy)-1,3-dimethyl-1H-pyrazole-4-carboxylic acid ≥95%
Supplier: Matrix Scientific
042928-500MG
042928-5G
101858-272EA
921.54
USD
101858-272
101858-276
5-(4-Chlorophenoxy)-1,3-dimethyl-1H-pyrazole-4-carboxylic acid ≥95%
5-(4-Chlorophenoxy)-1,3-dimethyl-1H-pyrazole-4-carboxylic acid
Hazard: Irritant
Formula:
C₁₂H₁₁ClN₂O₃ MW: 266.69 g/mol Melting Pt: 202…204 °C |
MDL Number:
MFCD00215391 CAS Number: 303149-69-1 |
Specification Test Results
Melting point | 202-204°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...