3-[(4-Fluorobenzyl)oxy]-2-thiophenecarboxylic acid ≥95%

Supplier: Matrix Scientific

042922-500MG 042922-5G
101858-238EA 375.38 USD
101858-238 101858-242
3-[(4-Fluorobenzyl)oxy]-2-thiophenecarboxylic acid ≥95%
3-[(4-Fluorobenzyl)oxy]-2-thiophenecarboxylic acid

Hazard: Irritant

Formula: C₁₂H₉FO₃S
Melting Pt: 118…120 °C
MDL Number: MFCD00173430
CAS Number: 303147-78-6

Order Now

Specification Test Results

Melting point 118-120°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR