Ethyl-N-(2-cyanoacetyl)carbamate ≥95%

Supplier: Matrix Scientific

Synonyms: N-Cyanoacetylurethane

042917-500MG 042917-5G
101858-210EA 32.31 USD
101858-210 101858-214
Ethyl-N-(2-cyanoacetyl)carbamate ≥95%
Ethyl-N-(2-cyanoacetyl)carbamate

Hazard: Irritant

Formula: C₆H₈N₂O₃
MW: 156.14 g/mol
Melting Pt: 167…169 °C
MDL Number: MFCD00001937
CAS Number: 6629-04-5

Order Now

Specification Test Results

Melting point 167-169°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR