Ethyl-2-cyano-2-(1,3-dioxolan-2-yliden)acetate ≥95%

Supplier: Matrix Scientific

042915-500MG 042915-5G
101858-198EA 278.46 USD
101858-198 101858-202
Ethyl-2-cyano-2-(1,3-dioxolan-2-yliden)acetate ≥95%
Ethyl-2-cyano-2-(1,3-dioxolan-2-yliden)acetate

Hazard: Irritant

Formula: C₈H₉NO₄
Melting Pt: 139…140 °C
MDL Number: MFCD00172357
CAS Number: 121020-70-0

Order Now

Specification Test Results

Melting point 139-140°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR