Methyl-1-oxo-2-(3-pyridinylmethyl)-1,2-dihydro-4-isoquinolinecarboxylate ≥95%

Supplier: Matrix Scientific

042906-500MG 042906-5G
101858-152EA 747.69 USD
101858-152 101858-156
Methyl-1-oxo-2-(3-pyridinylmethyl)-1,2-dihydro-4-isoquinolinecarboxylate ≥95%
Methyl-1-oxo-2-(3-pyridinylmethyl)-1,2-dihydro-4-isoquinolinecarboxylate

Hazard: Irritant

Formula: C₁₇H₁₄N₂O₃
MW: 294.31 g/mol
Melting Pt: 164…166 °C
MDL Number: MFCD00140334
CAS Number: 446276-03-5

Order Now

Specification Test Results

Melting point 164-166°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR