2-(1H-Imidazol-1-yl)benzenecarbonitrile ≥95%

Supplier: Matrix Scientific

042897-500MG 042897-5G
101858-104EA 181.54 USD
101858-104 101858-108
2-(1H-Imidazol-1-yl)benzenecarbonitrile ≥95%
2-(1H-Imidazol-1-yl)benzenecarbonitrile

Hazard: Irritant

Formula: C₁₀H₇N₃
Melting Pt: 146…148 °C
MDL Number: MFCD00127074
CAS Number: 25373-49-3

Order Now

Specification Test Results

Melting point 146-148°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR