5-Bromo-2-hydrazino-1H-1,3-benzimidazole ≥95%

Supplier: Matrix Scientific

042894-500MG 042894-5G
101858-088EA 375.38 USD
101858-088 101858-092
5-Bromo-2-hydrazino-1H-1,3-benzimidazole ≥95%
5-Bromo-2-hydrazino-1H-1,3-benzimidazole

Hazard: Irritant

Formula: C₇H₇BrN₄
MW: 227.07 g/mol
Melting Pt: 96…98 °C
MDL Number: MFCD08276936
CAS Number: 54303-63-8

Order Now

Specification Test Results

Melting point 96-98°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR