1-(4-Bromo-1-methyl-1H-pyrrol-2-yl)-2,2,2-trichloro-1-ethanone ≥95%
Supplier: Matrix Scientific
Synonyms:
4-Bromo-1-methyl-2-(trichloroacetyl)-1H-pyrrole
042889-500MG
042889-5G
101858-062EA
375.38
USD
101858-062
101858-066
1-(4-Bromo-1-methyl-1H-pyrrol-2-yl)-2,2,2-trichloro-1-ethanone ≥95%
1-(4-Bromo-1-methyl-1H-pyrrol-2-yl)-2,2,2-trichloro-1-ethanone
Hazard: Irritant
Formula:
C₇H₅BrCl₃NO MW: 305.39 g/mol Melting Pt: 102…104 °C |
MDL Number:
MFCD08276938 CAS Number: 184643-69-4 |
Specification Test Results
Melting point | 102-104°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...