2-[3-(Trifluoromethyl)phenyl]-1,3-thiazole-4-carbohydrazide ≥95%
Supplier: Matrix Scientific
042887-500MG
042887-5G
101858-050EA
556.92
USD
101858-050
101858-054
2-[3-(Trifluoromethyl)phenyl]-1,3-thiazole-4-carbohydrazide ≥95%
2-[3-(Trifluoromethyl)phenyl]-1,3-thiazole-4-carbohydrazide
Hazard: Irritant
Formula:
C₁₁H₈F₃N₃OS Melting Pt: 161…163 °C |
MDL Number:
MFCD01936330 CAS Number: 400083-64-9 |
Specification Test Results
Melting point | 161-163°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...