Ethyl-2-amino-5-bromonicotinate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-amino-5-bromonicotinate

042882-1G 042882-25G
101858-022EA 73.85 USD
101858-022 101858-026
Ethyl-2-amino-5-bromonicotinate ≥95%
Ethyl 2-amino-5-bromonicotinate

Hazard: Irritant

Formula: C₈H₉N₂O₂Br
MW: 245.0700 g/mol
Melting Pt: 139-141°C
MDL Number: MFCD08276943
CAS Number: 433226-06-3

Order Now

Specification Test Results

Melting point 139-141°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR