Methyl 1,6-dihydro-5-iodo-6-oxopyridine-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl-5-iodo-6-oxo-1,6-dihydro-3-pyridinecarboxylate, 1,2-Dihydro-3-iodo-5-(methoxycarbonyl)-2-oxopyridine, Methyl 1,6-dihydro-5-iodo-6-oxonicotinate

042877-500MG 042877-5G
101857-998EA 300 USD
101857-998 101858-002
Methyl 1,6-dihydro-5-iodo-6-oxopyridine-3-carboxylate ≥95%
Methyl 1,6-dihydro-5-iodo-6-oxopyridine-3-carboxylate

Hazard: Irritant

Formula: C₇H₆INO₃
MW: 279.04 g/mol
Melting Pt: 209…211 °C
MDL Number: MFCD08276965
CAS Number: 885950-46-9

Order Now

Specification Test Results

Melting point 209-211°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR