4'-Trifluoromethylbiphenyl-4-carbaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 4'-(Trifluoromethyl)-[1,1'-biphenyl]-4-carboxaldehyde, 4-(4-Trifluoromethylphenyl)benzaldehyde, 4'-Trifluoromethyl-biphenyl-4-carbaldehyde, 4'-(Trifluoromethyl)-[1,1'-biphenyl]-4-carbaldehyde

042875-500MG 042875-5G
101857-986EA 36.92 USD
101857-986 101857-990
4'-Trifluoromethylbiphenyl-4-carbaldehyde ≥95%
4'-Trifluoromethylbiphenyl-4-carbaldehyde

Hazard: Irritant

Formula: C₁₄H₉F₃O
MW: 250.2200 g/mol
Melting Pt: 70…72 °C
MDL Number: MFCD01862519
CAS Number: 90035-34-0

Order Now

Specification Test Results

Melting point 70-72°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR